APO455835185 Apollo Scientific Ltd

4-Iodo-alpha,alpha,beta,beta-tetrafluorophenetole

  • MDL number MFCD08458071
  • Cas Number : 863323-03-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Iodo-4-(1,1,2,2-tetrafluoroethoxy)benzene, 4-Iodophenyl 1,1,2,2-tetrafluoroethyl ether
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10353

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10353
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart