APO455835193 Apollo Scientific Ltd

2,4-Difluoroanisole

  • MDL number MFCD00010731
  • Cas Number : 452-10-8
  • Molecular Weight : 144.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.24
Flash Point 51
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 53/18mm
Product Link https://store.apolloscientific.co.uk/product/24-difluoroanisole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/24-difluoroanisole
pictograms GHS02;GHS07
hstatements H226;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart