APO455835194 Apollo Scientific Ltd

2,5-Difluoroanisole

  • MDL number MFCD00042561
  • Cas Number : 75626-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,4-Difluoro-2-methoxybenzene, 2,5-Difluorophenyl methyl ether
Purity 98
Flash Point 40
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 72-73/25mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1246

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1246
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart