APO455835195 Apollo Scientific Ltd

3,4-Difluoroanisole

  • MDL number MFCD00010730
  • Cas Number : 115144-40-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,2-Difluoro-4-methoxybenzene, 3,4-Difluorophenyl methyl ether
Purity 98
Density 1.216
Flash Point 55
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1248

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1248
pictograms GHS02
hstatements H226
Availability details for
Quantity
  • Availability
  • Add to Cart