APO455835211 Apollo Scientific Ltd

5-Bromo-2-fluorophenyl methyl sulphone

  • MDL number MFCD08458078
  • Cas Number : 914636-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-96
Synonym 4-Bromo-1-fluoro-2-(methylsulphonyl)benzene
Purity 99
Storage temp. Ambient Temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10716

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10716
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart