APO455835212 Apollo Scientific Ltd

2-Fluoro-5-nitrophenyl methyl sulphone

  • MDL number MFCD08458079
  • Cas Number : 914636-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128-130
Synonym 1-Fluoro-2-(methylsulphonyl)-4-nitrobenzene, 4-Fluoro-3-(methylsulphonyl)nitrobenzene
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10717

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10717
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart