APO455835217 Apollo Scientific Ltd

4'-Methoxy-3'-(trifluoromethyl)acetophenone

  • MDL number MFCD01091010
  • Cas Number : 149105-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56-61
Synonym 1-[4-Methoxy-3-(trifluoromethyl)phenyl]ethan-1-one, 5-Acetyl-2-methoxybenzotrifluoride, 4-Acetyl-2-(trifluoromethyl)anisole
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-methoxy-3-trifluoromethylacetophenone

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-methoxy-3-trifluoromethylacetophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart