APO455835221 Apollo Scientific Ltd

3-(Trifluoromethoxy)benzyl chloride

  • MDL number MFCD08458087
  • Cas Number : 89807-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Chloromethyl)-3-(trifluoromethoxy)benzene, 3-(Chloromethyl)phenyl trifluoromethyl ether
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10722

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10722
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart