APO455835237 Apollo Scientific Ltd

2-(Aminomethyl)-5-fluorobenzeneboronic acid, N-BOC protected

  • MDL number MFCD04115639
  • Cas Number : 850568-43-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98-102
Synonym 2-{[(tert-Butoxycarbonyl)amino]methyl}-5-fluorobenzeneboronic acid
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8843

Safety Information

signalword Warning
pcodes P280;P301+P312;P302+P352
MSDS http://www.apolloscientific.co.uk/display_item.php?id=8843
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart