APO455835239 Apollo Scientific Ltd

4-Ethoxy-3-fluorobenzeneboronic acid

  • MDL number MFCD04115667
  • Cas Number : 279263-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-113
Synonym 4-Borono-2-fluorophenetole
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-ethoxy-3-fluorobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/4-ethoxy-3-fluorobenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart