APO455835250 Apollo Scientific Ltd

2-Chloro-1-iodo-4-(trifluoromethoxy)benzene

  • MDL number MFCD08458089
  • Cas Number : 345226-19-9
  • Molecular Weight : 322.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Chloro-4-iodo-alpha,alpha,alpha-trifluoroanisole, 3-Chloro-4-iodophenyl trifluoromethyl ether
Purity 98
Storage temp. Store at 2-8
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-1-iodo-4-trifluoromethoxybenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-1-iodo-4-trifluoromethoxybenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart