APO455835253 Apollo Scientific Ltd

2,3'-Difluorobenzophenone

  • MDL number MFCD00017914
  • Cas Number : 58139-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2-Fluorophenyl)(3-fluorophenyl)methanone
Purity 97
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 142-143/3mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1293

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1293
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart