APO455835254 Apollo Scientific Ltd

2,4'-Difluorobenzophenone

  • MDL number MFCD00000319
  • Cas Number : 342-25-6
  • Molecular Weight : 218.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 22-24
Synonym (2-Fluorophenyl)(4-fluorophenyl)methanone
Purity 97
Density 1.244
Flash Point >110
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 176-178/16mm
Product Link https://store.apolloscientific.co.uk/product/24-difluorobenzophenone_PC2690B

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/24-difluorobenzophenone_PC2690B
pictograms GHS07;GHS09
hstatements H315;H319;H335;H411
Availability details for
Quantity
  • Availability
  • Add to Cart