APO455835256 Apollo Scientific Ltd

2,6-Difluorobenzophenone

  • MDL number MFCD00009895
  • Cas Number : 59189-51-4
  • Molecular Weight : 218.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2,6-Difluorophenyl)(phenyl)methanone
Purity 98
Density 1.244
Flash Point >110
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 120/2mm
Product Link https://store.apolloscientific.co.uk/product/26-difluorobenzophenone

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/26-difluorobenzophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart