APO455835264 Apollo Scientific Ltd

2,4,5-Trifluoroiodobenzene

  • MDL number MFCD04973898
  • Cas Number : 17533-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Iodo-2,4,5-trifluorobenzene
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/245-trifluoroiodobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/245-trifluoroiodobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart