APO455835265 Apollo Scientific Ltd

3,4,5-Trifluoroiodobenzene

  • MDL number MFCD03265259
  • Cas Number : 170112-66-0
  • Molecular Weight : 257.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Iodo-1,2,3-trifluorobenzene
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/345-trifluoroiodobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/345-trifluoroiodobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart