APO455835267 Apollo Scientific Ltd

2-Chloro-6-fluoroiodobenzene

  • MDL number MFCD08458092
  • Cas Number : 127654-70-0
  • Molecular Weight : 256.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Chloro-3-fluoro-2-iodobenzene
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-6-fluoroiodobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-6-fluoroiodobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart