APO455835272 Apollo Scientific Ltd

3,5-Bis(trifluoromethyl)phenylhydrazine hydrochloride

  • MDL number MFCD03094165
  • Cas Number : 502496-23-3
  • Molecular Weight : 280.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point ca 208(dec.)
Synonym 1,3-Bis(trifluoromethyl)-5-hydrazinobenzene hydrochloride
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/35-bistrifluoromethylphenylhydrazine-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/35-bistrifluoromethylphenylhydrazine-hydrochloride
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart