APO455835276 Apollo Scientific Ltd

3,4-Difluorophenylhydrazine hydrochloride

  • MDL number MFCD03094170
  • Cas Number : 875664-54-3
  • Molecular Weight : 180.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point ca 230(dec.)
Synonym 1,2-Difluoro-4-hydrazinobenzene hydrochloride
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/34-difluorophenylhydrazine-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/34-difluorophenylhydrazine-hydrochloride
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart