APO455835282 Apollo Scientific Ltd

2-Fluoro-4-iodobenzonitrile

  • MDL number MFCD03094172
  • Cas Number : 137553-42-5
  • Molecular Weight : 247.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp >110
bp 103-105
Iupac Name 2-fluoro-4-iodobenzonitrile
Inchi Code 1S/C7H3FIN/c8-7-3-6(9)2-1-5(7)4-10/h1-3H
Inchi Key WHQVXHBSTRFRCE-UHFFFAOYSA-N

Description

Melting Point >110
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 103-105
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-4-iodobenzonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-4-iodobenzonitrile
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart