APO455835287 Apollo Scientific Ltd

2,5-Difluorobenzyl alcohol

  • MDL number MFCD00010627
  • Cas Number : 75853-20-2
  • Molecular Weight : 144.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2,5-Difluorophenyl)methanol
Purity 98
Density 1.29
Flash Point 93
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/25-difluorobenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/25-difluorobenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart