APO455835290 Apollo Scientific Ltd

3,5-Difluorobenzyl alcohol

  • MDL number MFCD00010313
  • Cas Number : 79538-20-8
  • Molecular Weight : 144.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (3,5-Difluorophenyl)methanol
Purity 98
Density 1.276
Flash Point 94
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 98-99/9mm
Product Link https://store.apolloscientific.co.uk/product/35-difluorobenzyl-alcohol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/35-difluorobenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart