APO455835298 Apollo Scientific Ltd

2-Fluoro-4-iodobenzoic acid

  • MDL number MFCD03094173
  • Cas Number : 124700-40-9
  • Molecular Weight : 266.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 216-219
Iupac Name 2-fluoro-4-iodobenzoic acid
Inchi Code 1S/C7H4FIO2/c8-6-3-4(9)1-2-5(6)7(10)11/h1-3H,(H,10,11)
Inchi Key OLQSJAFBRCOGPQ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 216-219
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-4-iodobenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-4-iodobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart