APO455835305 Apollo Scientific Ltd

3,3'-Difluorobiphenyl

  • MDL number MFCD00039216
  • Cas Number : 396-64-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44050
Purity 98
Density 1.21
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 130/14mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1334

Safety Information

signalword Warning
pcodes P261;P280;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1334
pictograms GHS07
hstatements H315;H319;H335;H412
Availability details for
Quantity
  • Availability
  • Add to Cart