APO455835319 Apollo Scientific Ltd

3-Fluoro-5-methoxybenzaldehyde

  • MDL number MFCD08741376
  • Cas Number : 699016-24-5
  • Molecular Weight : 154.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 24-29
Synonym 5-Fluoro-m-anisaldehyde, 3-Fluoro-5-formylanisole
Purity 97
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-5-methoxybenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-5-methoxybenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart