APO455835321 Apollo Scientific Ltd

4,4'-Difluorodiphenylmethane

  • MDL number MFCD00000360
  • Cas Number : 457-68-1
  • Molecular Weight : 204.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 29-30
Synonym 1,1'-Methylenebis(4-fluorobenzene), 1,1'-Methanediylbis(4-fluorobenzene)
Purity 99
Density 1.145
Flash Point >110
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 156-160/12mm
Product Link https://store.apolloscientific.co.uk/product/44-difluorodiphenylmethane

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/44-difluorodiphenylmethane
pictograms GHS07
hstatements H312;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart