APO455835324 Apollo Scientific Ltd

4,5-Difluoro-2-nitrophenol

  • MDL number MFCD03094187
  • Cas Number : 55346-97-9
  • Molecular Weight : 175.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54-57
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/45-difluoro-2-nitrophenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/45-difluoro-2-nitrophenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart