APO455835328 Apollo Scientific Ltd

4-Bromo-2-fluoro-6-nitrotoluene

  • MDL number MFCD03094190
  • Cas Number : 502496-34-6
  • Molecular Weight : 234.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-3-fluoro-2-methylnitrobenzene
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-fluoro-6-nitrotoluene

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-fluoro-6-nitrotoluene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart