APO455835334 Apollo Scientific Ltd

3-Fluoro-5-methoxybenzyl alcohol

  • MDL number MFCD08741377
  • Cas Number : 914637-27-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 27-30
Synonym (3-Fluoro-5-methoxyphenyl)methanol, 3-Fluoro-5-(hydroxymethyl)anisole
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-5-methoxybenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-5-methoxybenzyl-alcohol
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart