APO455835338 Apollo Scientific Ltd

2,2-Difluoroethan-1-ol

  • MDL number MFCD00042248
  • Cas Number : 359-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -28
Synonym 1,1-Difluoro-2-hydroxyethane
Purity 97
Density 1.296
Flash Point 3
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 95-96
Product Link https://store.apolloscientific.co.uk/product/22-difluoroethan-1-ol-97

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/22-difluoroethan-1-ol-97
pictograms GHS02;GHS07
hstatements H225;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart