APO455835341 Apollo Scientific Ltd

5-Chloro-2-nitro-4-(trifluoromethoxy)aniline

  • MDL number MFCD04972735
  • Cas Number : 914637-23-3
  • Molecular Weight : 256.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-chloro-2-nitro-4-trifluoromethoxyaniline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-2-nitro-4-trifluoromethoxyaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart