APO455835351 Apollo Scientific Ltd

2,4-Difluorophenacylamine hydrochloride

  • MDL number MFCD07783701
  • Cas Number : 786719-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Amino-2',4'-difluoroacetophenone hydrochloride
Purity 99
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=11081

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=11081
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart