APO455835354 Apollo Scientific Ltd

2,4-Difluoromandelic acid

  • MDL number MFCD00061296
  • Cas Number : 132741-30-1
  • Molecular Weight : 188.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122-124
Synonym alpha-Hydroxy-2,4-difluorophenylacetic acid
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-difluoromandelic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/24-difluoromandelic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart