APO455835366 Apollo Scientific Ltd

2,4-Difluoro-6-nitroaniline

  • MDL number MFCD00034058
  • Cas Number : 364-30-7
  • Molecular Weight : 174.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85-87
Purity 98
Flash Point >110
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-difluoro-6-nitroaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/24-difluoro-6-nitroaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart