APO455835368 Apollo Scientific Ltd

2-Fluoro-3-nitrotoluene

  • MDL number MFCD03412242
  • Cas Number : 437-86-5
  • Molecular Weight : 155.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 18-19
Synonym 2-Fluoro-3-methylnitrobenzene, 2-Fluoro-1-methyl-3-nitrobenzene
Density 1.28
Flash Point 98
Storage temp. Store at 2-8
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 110-111/12mm
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-3-nitrotoluene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-3-nitrotoluene
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart