APO455835370 Apollo Scientific Ltd

2-Bromo-6-(trifluoromethyl)benzoic acid

  • MDL number MFCD06253757
  • Cas Number : 177420-64-3
  • Molecular Weight : 269.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122-128
Synonym 3-Bromo-2-carboxybenzotrifluoride, 6-Bromo-alpha,alpha,alpha-trifluoro-o-toluic acid
Purity 97
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-6-trifluoromethylbenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-6-trifluoromethylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart