APO455835390 Apollo Scientific Ltd

2,3-Difluorophenylacetonitrile

  • MDL number MFCD00061277
  • Cas Number : 145689-34-5
  • Molecular Weight : 153.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,3-Difluorobenzyl cyanide
Purity 97
Density 1.2
Flash Point 85
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/23-difluorophenylacetonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/23-difluorophenylacetonitrile
pictograms GHS07
hstatements H302;H312;H319;H332
Availability details for
Quantity
  • Availability
  • Add to Cart