APO455835400 Apollo Scientific Ltd

2,4-Difluorophenylhydrazine

  • MDL number MFCD00042256
  • Cas Number : 40594-30-7
  • Molecular Weight : 144.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65-67
Synonym 2,4-Difluoro-1-hydrazinobenzene
Purity 95
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-difluorophenylhydrazine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/24-difluorophenylhydrazine
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart