APO455835402 Apollo Scientific Ltd

2,5-Difluorophenylhydrazine hydrochloride

  • MDL number MFCD00013385
  • Cas Number : 175135-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210-211(dec.)
Synonym 1,4-Difluoro-2-hydrazinobenzene hydrochloride
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1455

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1455
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart