APO455835411 Apollo Scientific Ltd

2',3'-Difluoropropiophenone

  • MDL number MFCD00061141
  • Cas Number : 236754-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(2,3-Difluorophenyl)propan-1-one
Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/23-difluoropropiophenone

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/23-difluoropropiophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart