APO455835413 Apollo Scientific Ltd

2',6'-Difluoropropiophenone

  • MDL number MFCD00015506
  • Cas Number : 85068-31-1
  • Molecular Weight : 170.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2,6-difluorophenyl)-1-propanone
Inchi Code 1S/C9H8F2O/c1-2-8(12)9-6(10)4-3-5-7(9)11/h3-5H,2H2,1H3
Inchi Key ISFKUAKHXQLAFN-UHFFFAOYSA-N

Description

Synonym 1-(2,6-Difluorophenyl)propan-1-one
Flash Point 70-71/4mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-difluoropropiophenone

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/26-difluoropropiophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart