APO455835414 Apollo Scientific Ltd

3',5'-Difluoropropiophenone

  • MDL number MFCD00061142
  • Cas Number : 135306-45-5
  • Molecular Weight : 170.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 25-27
Iupac Name 1-(3,5-difluorophenyl)-1-propanone
Inchi Code 1S/C9H8F2O/c1-2-9(12)6-3-7(10)5-8(11)4-6/h3-5H,2H2,1H3
Inchi Key FVDQWXARVQADKN-UHFFFAOYSA-N

Description

Purity 97%
Flash Point 82/10mm
Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 25-27
Product Link https://store.apolloscientific.co.uk/product/35-difluoropropiophenone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/35-difluoropropiophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart