APO455835416 Apollo Scientific Ltd

2,2-Difluorosuccinic acid

  • MDL number MFCD00040970
  • Cas Number : 665-31-6
  • Molecular Weight : 154.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 144-145
Iupac Name 2,2-difluorosuccinic acid
Inchi Code 1S/C4H4F2O4/c5-4(6,3(9)10)1-2(7)8/h1H2,(H,7,8)(H,9,10)
Inchi Key ZYLFHISLYSHWRH-UHFFFAOYSA-N

Description

Synonym 2,2-Difluorobutane-1,4-dioic acid
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 144-145
Product Link https://store.apolloscientific.co.uk/product/22-difluorosuccinic-acid

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/22-difluorosuccinic-acid
pictograms GHS05;GHS07
hstatements H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart