APO455835422 Apollo Scientific Ltd

2-Bromo-4-fluoronitrobenzene

  • MDL number MFCD00792441
  • Cas Number : 700-36-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 39-42
Synonym 2-Bromo-4-fluoro-1-nitrobenzene
Purity 98
Flash Point >110
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 70-75/0.4mm
Product Link https://store.apolloscientific.co.uk/product/2-bromo-4-fluoronitrobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-4-fluoronitrobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart