APO455835429 Apollo Scientific Ltd

5-Fluoro-3-(piperidin-4-yl)-1,2-benzisoxazole hydrochloride

  • MDL number MFCD06200879
  • Cas Number : 84163-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(5-Fluoro-1,2-benzisoxazol-3-yl)piperidine hydrochloride
Purity 97
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=11129

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=11129
Availability details for
Quantity
  • Availability
  • Add to Cart