APO455835431 Apollo Scientific Ltd

1-(Trifluoromethyl)ethylamine hydrochloride

  • MDL number MFCD04972986
  • Cas Number : 2968-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218-221(subl.)
Synonym 1,1,1-Trifluoropropan-2-amine hydrochloride, 2-Amino-1,1,1-trifluoropropane hydrochloride
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/1-trifluoromethylethylamine-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-trifluoromethylethylamine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart