APO455835433 Apollo Scientific Ltd

5-Bromo-2,3-diaminobenzotrifluoride

  • MDL number MFCD08741393
  • Cas Number : 157026-19-2
  • Molecular Weight : 255.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-3-(trifluoromethyl)benzene-1,2-diamine, 5-Bromo-1,2-diamino-3-(trifluoromethyl)benzene
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-23-diaminobenzotrifluoride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-23-diaminobenzotrifluoride
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart