APO455835445 Apollo Scientific Ltd

3-Hydroxy-5-nitrobenzotrifluoride

  • MDL number MFCD04973779
  • Cas Number : 349-57-5
  • Molecular Weight : 207.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89-92
Synonym 3-Nitro-5-(trifluoromethyl)phenol, 3-Hydroxy-5-(trifluoromethyl)nitrobenzene
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-hydroxy-5-nitrobenzotrifluoride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-hydroxy-5-nitrobenzotrifluoride
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart