APO455835460 Apollo Scientific Ltd

5-Bromo-2-fluoro-3-methylpyridine

  • MDL number MFCD03095091
  • Cas Number : 29312-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-65
Synonym 5-Bromo-2-fluoro-3-picoline
Density 1.6
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-fluoro-3-methylpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-fluoro-3-methylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart