APO455835462 Apollo Scientific Ltd

4-Acetyl-3-fluorobenzeneboronic acid

  • MDL number MFCD06796422
  • Cas Number : 481725-35-3
  • Molecular Weight : 181.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4'-Borono-2'-fluoroacetophenone
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-acetyl-3-fluorobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-acetyl-3-fluorobenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart